4-Biphenylmethyl chloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Chloromethyl)biphenyl

OR6777-1G OR6777-25G OR6777-5G
76595-120EA 20.87 USD
76595-120 76595-124 76595-122
4-Biphenylmethyl chloride
4-Biphenylmethyl chloride
Formula: C₁₃H₁₁Cl
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00000917
CAS Number: 1667-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR