4-Chloro-3-nitrobenzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR6710-1G OR6710-5G
76621-826EA 20.87 USD
76621-826 76621-828
4-Chloro-3-nitrobenzoyl chloride
4-Chloro-3-nitrobenzoyl chloride
Formula: C₇H₃Cl₂NO₃
Melting Pt: 47…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00035743
CAS Number: 38818-50-7
EINECS: 254-133-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR