(R)-(+)-1,1'-Bi-2-naphthol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-BINOL(R)-(+)-1,1'-Binaphthyl-2,2'-Diol(R)-[1,1'-Binaphthalene]-2,2'-diol

OR6692-250G OR6692-100G OR6692-10G
76617-214EA 32.96 USD
76617-214 76617-216 76617-212
(R)-(+)-1,1'-Bi-2-naphthol 97%
(R)-(+)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
MW: 286.33 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 18531-94-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR