ω,3-Dichloroacetophenone

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Chlorophenacyl chloride

OR6674-1G OR6674-25G OR6674-5G
76621-810EA 63.16 USD
76621-810 76621-812 76621-814
ω,3-Dichloroacetophenone
ω,3-Dichloroacetophenone
Formula: C₈H₆Cl₂O
Melting Pt: 40…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783975
CAS Number: 21886-56-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR