2-Iodobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR6669-1G OR6669-5G
76595-028EA 28.52 USD
76595-028 76595-030
2-Iodobenzaldehyde
2-Iodobenzaldehyde
Formula: C₇H₅IO
Boiling Pt: 102 °C/3mmHg
Melting Pt: 36…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS Number: 26260-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR