4-Amino-m-anisic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

OR6621-100G OR6621-25G OR6621-5G
76594-988EA 288.28 USD
76594-988 76594-986 76594-984
4-Amino-m-anisic acid
4-Amino-m-anisic acid
Formula: C₈H₉NO₃
MW: 167.16
Melting Pt: 186…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016539
CAS Number: 2486-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR