Indoline 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Dihydroindol

OR6520-250G OR6520-25G
76594-858EA 147.72 USD
76594-858 76594-856
Indoline 98%
Indoline
Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220
Melting Pt: -21
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR