N,N-Dimethylethylendiamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

OR6514-100G OR6514-250G
76612-288EA 20.38 USD
76612-288 76612-290 76612-292
N,N-Dimethylethylendiamine 97%
N,N-Dimethylethylendiamine
Formula: C₄H₁₂N₂
Boiling Pt: 107 °C
Density: 0.81 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2
UN: 2733
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR