Imidazo[1,2-b]pyridazine-2-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR6356-1G OR6356-250MG
76594-714EA 116.12 USD
76594-714 76594-712
Imidazo[1,2-b]pyridazine-2-carboxylic acid 97%
Imidazo[1,2-b]pyridazine-2-carboxylic acid
Formula: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08235229
CAS Number: 160911-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR