Furan-2,5-dicarboxylic acid 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,5-Furandicarboxylic acid
OR6215-25G
OR6215-5G
OR6215-100G
76594-534EA
70
USD
76594-534
76594-532
76594-536
Furan-2,5-dicarboxylic acid 97%
Furan-2,5-dicarboxylic acid
Formula:
C₆H₄O₅ MW: 156.09 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016582 CAS Number: 3238-40-2 EINECS: 221-800-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...