4-Aminomethylbenzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (4-Aminomethylphenyl)methanol, (4-(Aminomethyl)phenyl)methanol, 4-Aminomethylbenzyl alcohol, 4-(Aminomethyl)benzyl alcohol

OR6193-1G OR6193-250MG OR6193-5G
76617-174EA 55.02 USD
76617-174 76617-176 76617-178
4-Aminomethylbenzyl alcohol 95%
4-Aminomethylbenzyl alcohol
Formula: C₈H₁₁NO
Melting Pt: 82…83 °C
Storage Temperature: Ambient
MDL Number: MFCD06213766
CAS Number: 39895-56-2
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR