2,1,3-Benzothiadiazol-4-ylamine 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR6147-1G OR6147-5G
76617-152EA 41.76 USD
76617-152 76617-154
2,1,3-Benzothiadiazol-4-ylamine 95+%
2,1,3-Benzothiadiazol-4-ylamine
Formula: C₆H₅N₃S
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS Number: 767-64-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR