3-Aminopyridazine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pyridazin-3-amine

OR6130-1G OR6130-5G
76594-208EA 20.26 USD
76594-208 76594-210
3-Aminopyridazine 97%
3-Aminopyridazine
Formula: C4H5N3
MW: 95.10 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD01529869
CAS Number: 5469-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR