2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
DavePhos
, 2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine
, 2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl
, 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl
OR61059-5G
OR61059-25G
76594-024EA
30.56
USD
76594-024
76594-026
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl
Usage subject to US Patents 6307087 and 6395916. This product is intended for research and development purposes only.
Formula:
C₂₆H₃₆NP MW: 393.55 g/mol Melting Pt: 121…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD02183572 CAS Number: 213697-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...