2-Amino-4,6-dichloropyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR60009-10G OR60009-1G OR60009-5G
76593-588EA 135.48 USD
76593-588 76593-584 76593-586
2-Amino-4,6-dichloropyridine
2-Amino-4,6-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 108…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037218
CAS Number: 116632-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR