Glyoxal dimethyl acetal 60% in aqueous solution

Supplier: APOLLO SCIENTIFIC
Warning

OR59985-100G OR59985-500G OR59985-5G
76593-566EA 75.4 USD
76593-566 76593-568 76593-564
Glyoxal dimethyl acetal 60% in aqueous solution
Glyoxal dimethyl acetal
Formula: OHCCH(OCH₃)₂
Boiling Pt: 100…102 °C/759mmHg
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00134410
CAS Number: 51673-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR