2-Thiophenemethanol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Thiophen-2-ylmethanolThiophen-2-methanol2-Thienylmethanol

OR59889-100G OR59889-500G OR59889-25G
76593-486EA 85 USD
76593-486 76593-488 76593-484
2-Thiophenemethanol 99%
2-Thiophenemethanol
Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 206…207 °C
Density: 1.205 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005454
CAS Number: 636-72-6
EINECS: 211-264-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR