5-Bromo-1,3,4-thiadiazol-2-amine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-5-bromo-1,3,4-thiadiazole, 1,3,4-Thiadiazol-2-amine, 5-bromo-, 5-bromo-1,3,4-thiadiazol-2-amine

OR59876-25G OR59876-5G
76593-476EA 157.59 USD
76593-476 76593-474 76593-472
5-Bromo-1,3,4-thiadiazol-2-amine
5-Bromo-1,3,4-thiadiazol-2-amine
Formula: C₂H₂BrN₃S
Melting Pt: 178…182 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00464325
CAS Number: 37566-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR