2-Amino-3-bromopyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR59810-10G OR59810-25G OR59810-5G
76593-388EA 34.64 USD
76593-388 76593-390 76593-386
2-Amino-3-bromopyridine 98%
2-Amino-3-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03095187
CAS Number: 13534-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR