Azetidine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR59419-5G
OR59419-25G
OR59419-1G
76612-156EA
124
USD
76612-156
76612-154
76612-152
Azetidine
Azetidine
Formula:
C₃H₇N Boiling Pt: 61…62 °C Density: 0.847 g/cm³ (25 °C) Flash Pt: –21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005165 EINECS: 207-963-8 UN: 2733 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...