5-Amino-2-methylindole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole

OR59321-100G OR59321-25G
76593-216EA 206.78 USD
76593-216 76593-218 76593-214
5-Amino-2-methylindole 95%
5-Amino-2-methylindole
Formula: C₉H₁₀N₂
MW: 146.19 g/mol
Melting Pt: 153…157 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093426
CAS Number: 7570-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR