D-(-)-N-Methylglucamine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
OR5855-250G
OR5855-50G
76593-156EA
55.02
USD
76593-156
76593-154
D-(-)-N-Methylglucamine
D(-)-N-Methylglucamine
Formula:
H₃CNHCH₂(CH(OH))₄CH₂OH Melting Pt: 129…131.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004707 CAS Number: 6284-40-8 EINECS: 228-506-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...