Methyl-3-amino-2-methylbenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester
OR5797-100G
OR5797-25G
76593-060EA
80.48
USD
76593-060
76593-058
Methyl-3-amino-2-methylbenzoate
Methyl 3-amino-2-methylbenzoate
Formula:
C₉H₁₁NO₂ Boiling Pt: 300 °C Density: 1.146 g/cm³ (25 °C) Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211328 CAS Number: 18583-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...