2-Amino-6-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5749-100G OR5749-25G
76593-016EA 85.58 USD
76593-016 76593-014
2-Amino-6-chloropyridine 98%
2-Amino-6-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR