3-Amino-5-tert-butylisoxazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR5616-25G OR5616-5G
76592-860EA 277.07 USD
76592-860 76592-858 76592-856
3-Amino-5-tert-butylisoxazole 97%
3-Amino-5-tert-butylisoxazole
Formula: C₇H₁₂N₂O
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00055620
CAS Number: 55809-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR