3-Aminoisoxazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR5608-1G OR5608-5G
76592-828EA 20.87 USD
76592-828 76592-830
3-Aminoisoxazole 95%
3-Aminoisoxazole
Formula: C₃H₄N₂O
Boiling Pt: 226…228 °C
Density: 1.138 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00038814
CAS Number: 1750-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR