2-Amino-4-(4-bromophenyl)thiazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5603-25G OR5603-5G OR5603-1G
76592-826EA 83.52 USD
76592-826 76592-824 77423-156
2-Amino-4-(4-bromophenyl)thiazole 98%
2-Amino-4-(4-bromophenyl)thiazole
Formula: C₉H₇BrN₂S
Melting Pt: 181…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051953
CAS Number: 2103-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR