Ethyl-2-aminoisonicotinate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

OR5579-10G OR5579-1G OR5579-5G
76592-784EA 34.38 USD
76592-784 76592-780 76592-782
Ethyl-2-aminoisonicotinate 97%
Ethyl 2-aminoisonicotinate
Formula: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03791260
CAS Number: 13362-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR