5-Aminopentan-1-ol, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR55441-100G OR55441-25G OR55441-500G
76617-012EA 140.14 USD
76617-012 76617-014 76617-016
5-Aminopentan-1-ol, Technical Grade
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR