8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR55437-1G OR55437-250MG
76592-502EA 956.52 USD
76592-502 76592-500
8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
8-Nitro-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide
Formula: C₁₀H₁₀N₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD31629919
CAS Number: 2169311-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR