1-Bromo-2-butyne 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5530-100G OR5530-25G OR5530-5G
76612-100EA 90.68 USD
76612-100 76612-102 76612-104
1-Bromo-2-butyne 98%
1-Bromo-2-butyne
Formula: C₄H₅Br
Boiling Pt: 123…125 °C
Density: 1.519 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR