3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonateCHAPS

61195-0050 61195-0010
TS61195-0050EA 184 USD
TS61195-0050 TS61195-0010
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00081083
CAS Number: 75621-03-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility (1 %) Clear colorless
TLC ≥98 %
UV (1 %)
UV at 260 nm A: ≤0.1
UV at 280 nm A: ≤0.1
Identification Passes test
pH 5.0 to 7.0 (10 % solution)
Conductivity ≤50 µmho/cm (10% aq. solution, 24°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR