Light green SF yellowish, certified by the Biological Stain Commission

Supplier: Thermo Scientific Chemicals

Synonyms: CI 42095Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonateDihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium saltSodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonateacid Green FLight Green 2G, S or 2GNDihydrogen (ethyl)[4-[p-[ethyl(m-sulfonatobenzyl)amino]-p'-sulfonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulfonatobenzyl)ammonium disodium saltacid Green 5Fast acid Green NCN DcL-30Food Green n°2

61185-0250
TS61185-0250EA 111 USD
TS61185-0250
Light green SF yellowish, certified by the Biological Stain Commission
Light Green SF Yellowish
Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
Melting Pt: 288…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00012121
CAS Number: 5141-20-8

Order Now

Specification Test Results

Additional info BSC certification: passes test
Dye content report

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR