Dextran sulfate sodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
Dextran sulfate sodium from Dextran
, Dextran sulphate sodium from Dextran
61136-1000
TS61136-1000EA
271
USD
TS61136-1000
Dextran sulfate sodium salt
Dextran sulphate sodium salt
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Ambient |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Specification Test Results
Appearance | White to off-white Powder |
Loss on drying | ≤7.5 % () |
Additional info | sulfur: 17 - 20 % |
Identification | passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...