Trimethylamine-N-oxide dihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylamine oxide dihydrate

42189-0250 42189-1000 42189-5000
TS42189-0250EA 102 USD
TS42189-0250 TS42189-1000 TS42189-5000
Trimethylamine-N-oxide dihydrate 98%
Trimethylamine N-oxide dihydrate
Formula: C₃H₉NO·2H₂O
Melting Pt: 94…98 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00149077
CAS Number: 62637-93-8
EINECS: 214-675-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 94 to 98 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR