Triethanolamine (trolamine) 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2',2''-NitrilotriethanolTris-(2-hydroxyethyl)amine

42163-0250 42163-0025
TS42163-0250EA 1456 USD
TS42163-0250 TS42163-0025
Triethanolamine (trolamine) 99+%
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 21 °C
Density: 1.1245 g/cm³
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20 °C, 589 nm)
Titration with HClO4 ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR