(S)-(+)-Prolinol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol

41832-0010 41832-0050 41832-0250
TS41832-0010EA 31 USD
TS41832-0010 TS41832-0050 TS41832-0250
(S)-(+)-Prolinol 95%
(S)-(+)-Prolinol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 74…76 °C (2 mmHg)
Density: 1.036 g/cm³
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005255
CAS Number: 23356-96-9
EINECS: 245-605-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4840 to 1.4870 (20 °C, 589 nm)
GC ≥94 %
Specific optical rotation +29° to +33° (20 °C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR