L(+)-Penicillamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine

41687-0010 41687-0050
TS41687-0010EA 55 USD
TS41687-0010 TS41687-0050
L(+)-Penicillamine 98%
L(+)-Penicillamine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: 206 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00064303
CAS Number: 1113-41-3
EINECS: 214-203-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation +61° to +65° (20 °C, 589 nm) (c=1, 1 M NaOH)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR