1,6-Diaminohexane 60% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

41156-0010 41156-2500
TS41156-0010EA 96 USD
TS41156-0010 TS41156-2500
1,6-Diaminohexane 60% (w/w) in aqueous solution
1,6-Diaminohexane
Formula: C₆H₁₆N₂
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 1783
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤20 APHA
Strength 59 to 61 wt%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR