Geraniol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(E)-3,7-Dimethylocta-2,6-dien-1-ol
41090-1000
41090-0025
41090-5000
41090-0050
41090-0250
TS41090-0050EA
21.5
USD
TS41090-0050
TS41090-5000
TS41090-0250
TS41090-1000
TS41090-0025
Geraniol 99%
Geraniol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 229…230 °C (757 mmHg) Melting Pt: –15 °C Density: 0.878 g/cm³ Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002917 CAS Number: 106-24-1 EINECS: 203-377-1 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4750 to 1.4780 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...