2-Aminoterephthalic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminoterephthalic Acid

27803-0250 27803-1000
TS27803-0250EA 86 USD
TS27803-0250 TS27803-1000
2-Aminoterephthalic acid 99%
2-Aminoterephthalic acid
Formula: C₈H₇NO₄
Melting Pt: 324 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00134536
CAS Number: 10312-55-7

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR