Arecolinium bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

25013-1000 25013-0050 25013-0250
TS25013-1000EA 479 USD
TS25013-1000 TS25013-0050 TS25013-0250
Arecolinium bromide 98+%
Arecolinium bromide
Formula: C₈H₁₄BrNO₂
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00039041
CAS Number: 300-08-3
EINECS: 206-087-3
UN: 1544
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 168 to 174 °C
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR