L(-)-Carnitine 99+%

Supplier: Thermo Scientific Chemicals
Warning

24104-0010 24104-0100 24104-0500
TS24104-0010EA 49 USD
TS24104-0010 TS24104-0100 TS24104-0500
L(-)-Carnitine 99+%
L(-)-Carnitine
Formula: C₇H₁₅NO₃
Melting Pt: 208…212 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Specific optical rotation -29° to -32° (20 °C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR