Linalyl acetate (synthetic) 95%

Supplier: Thermo Scientific Chemicals
Warning

23230-0050 23230-1000 23230-5000
TS23230-0050EA 21 USD
TS23230-0050 TS23230-1000 TS23230-5000
Linalyl acetate (synthetic) 95%
Linalyl acetate
Formula: C₁₂H₂₀O₂
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00008907
CAS Number: 115-95-7
EINECS: 204-116-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20 °C, 589 nm)
GC ≥94 %
Specific gravity (20 °C) 0.898 to 0.903


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR