Creatine (N-amidinosarcosine) monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

22679-0250 22679-1000 22679-5000
TS22679-0250EA 28 USD
TS22679-0250 TS22679-1000 TS22679-5000
Creatine (N-amidinosarcosine) monohydrate 99%
Creatine (N-Amidinosarcosine) monohydrate
Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
Melting Pt: 292 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤12.6 % (105 °C)
Solubility (1 % in water) Clear colorless to light yellow
HPLC ≥98.5 % (on dry substance)
Residue after ignition ≤1 %
Titration with HClO4 ≥98.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR