D-(+)-Glucurono-6,3-lactone 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone
22592-0250
22592-1000
22592-5000
TS22592-0250EA
29
USD
TS22592-0250
TS22592-1000
TS22592-5000
D-(+)-Glucurono-6,3-lactone 99+%
D(+)-Glucurono-6,3-lactone
Formula:
C₆H₈O₆ Melting Pt: 170…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00135622 CAS Number: 32449-92-6 EINECS: 251-053-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 170 to 176 °C |
Loss on drying | ≤0.5 % (105 °C, 3 h) |
Solubility | (5 % in water) Clear colorless to light yellow |
Specific optical rotation | +17.0° to +20.0° (20 °C, 589 nm) (c=1, water) |
Titration excess NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...