(S)-(+)-2-Aminobutyric acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-2-Aminobutyric acidL-alpha-Amino-N-Butyric acid(S)-2-Aminobutanoic acidH-Abu-OH

21538-0050 21538-0010
TS21538-0050EA 236 USD
TS21538-0050 TS21538-0010
(S)-(+)-2-Aminobutyric acid 98%
(S)-(+)-2-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064415
CAS Number: 1492-24-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.1 %
Specific optical rotation +20.2° to +23° (20 °C, 589 nm) (c=2, 5 N HCl)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR