Thiocarbohydrazide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TCH

20753-0050 20753-0250 20753-1000
TS20753-1000EA 475 USD
TS20753-1000 TS20753-0250 TS20753-0050
Thiocarbohydrazide 98%
Thiocarbohydrazide
Formula: CH₆N₄S
MW: 106.15 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00007616
CAS Number: 2231-57-4
EINECS: 218-769-8
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White to beige to grey Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C (Dec)
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR