Dimethyl-2-aminoterephthalate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

19478-0500 19478-0250
TS19478-0500EA 84 USD
TS19478-0500 TS19478-0250
Dimethyl-2-aminoterephthalate 99%
Dimethyl 2-aminoterephthalate
Formula: C₁₀H₁₁NO₄
Melting Pt: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00008427
CAS Number: 5372-81-6
EINECS: 226-364-2

Order Now

Specification Test Results

Appearance Yellow to beige to light green to brown Powder or crystals
Infrared spectrum Conforms
Melting point 131 to 134 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR