N-Isopropylcyclohexylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

17176-1000 17176-0050 17176-0250
TS17176-1000EA 318 USD
TS17176-1000 TS17176-0050 TS17176-0250
N-Isopropylcyclohexylamine 99%
N-Isopropylcyclohexylamine
Formula: C₉H₁₉N
Density: 0.85 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00003831
CAS Number: 1195-42-2
EINECS: 214-798-5
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20 °C, 589 nm)
Titration with HClO4 ≥98.5 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR